Learn More
Medchemexpress LLC Fenipentol | 583-03-9 | 98.3% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB1273100MG
Encompass_Preferred
Fenipentol (1-Phenyl-1-pentanol) is a synthetic derivative of an ingredient found in Curcuma longa, which is utilized as both a condiment and a dye. This compound acts as an orally active choleretic agent, significantly influencing the release of secretin and gastrin, as well as contributing to the pancreatic secretion of bicarbonate and protein.
  • Synthetic derivative of an ingredient of Curcuma longa.
  • Functions as an orally active choleretic agent.
  • Plays a role in the release of secretin.
  • Plays a role in the release of gastrin.
  • Important for pancreatic secretion of bicarbonate.
  • Important for pancreatic secretion of protein.
Catalog No. 50-004-13016