Learn More
Medchemexpress LLC Fenitrothion | 122-14-5 | 99.4% | 100 MG
SDP
Catalog No. 5000409639
Encompass_Preferred
Fenitrothion is a broad-spectrum, orally active insecticide and acaricide. It functions by inhibiting cholinesterase, AMPKα, and IRS1/PI3K/AKT. This compound also induces apoptosis and reduces SOD activity, making it effective against pests such as *Rhyzopertha dominica* and *Tribolium castaneum* adults.
  • Inhibits cholinesterase, AMPKα, and IRS1/PI3K/AKT.
  • Induces apoptosis and reduces SOD activity.
  • Effective against *Rhyzopertha dominica* and *Tribolium castaneum* adults.
  • Widely used in cotton, vegetable, fruit, and field crops, particularly rice.
  • Can be utilized for brain and spleen toxicology studies.
  • For research use only.
Catalog No. 50-004-09639 Supplier Medchemexpress LLC Supplier No. HYB1885100MG
Fenitrothion is a broad-spectrum, orally active insecticide and acaricide. It functions by inhibiting cholinesterase, AMPKα, and IRS1/PI3K/AKT. This compound also induces apoptosis and reduces SOD activity, making it effective against pests such as *Rhyzopertha dominica* and *Tribolium castaneum* adults.
  • Inhibits cholinesterase, AMPKα, and IRS1/PI3K/AKT.
  • Induces apoptosis and reduces SOD activity.
  • Effective against *Rhyzopertha dominica* and *Tribolium castaneum* adults.
  • Widely used in cotton, vegetable, fruit, and field crops, particularly rice.
  • Can be utilized for brain and spleen toxicology studies.
  • For research use only.