Learn More
Medchemexpress LLC Fenitrothion | 122-14-5 | 99.43% | 250 MG
SDP
Catalog No. 5000414339
Encompass_Preferred
Fenitrothion is a broad-spectrum and orally active insecticide/acaricide. It functions by inhibiting cholinesterase, AMPKα, and IRS1/PI3K/AKT, and it can also cause apoptosis and reduce SOD activity. This compound is widely utilized in various crops such as cotton, vegetables, fruits, and field crops, particularly rice.
  • Demonstrates insecticidal effects against adults of *Rhyzopertha dominica* and *Tribolium castaneum*.
  • Can be employed in brain and spleen toxicology studies.
  • Induces glucose metabolism disorders in rat liver BRL cells.
  • Selectively affects splenic T cells in Wistar rats.
Catalog No. 50-004-14339 Supplier Medchemexpress LLC Supplier No. 250MGHYB1885
Add to Cart
Add to Cart
Fenitrothion is a broad-spectrum and orally active insecticide/acaricide. It functions by inhibiting cholinesterase, AMPKα, and IRS1/PI3K/AKT, and it can also cause apoptosis and reduce SOD activity. This compound is widely utilized in various crops such as cotton, vegetables, fruits, and field crops, particularly rice.
  • Demonstrates insecticidal effects against adults of *Rhyzopertha dominica* and *Tribolium castaneum*.
  • Can be employed in brain and spleen toxicology studies.
  • Induces glucose metabolism disorders in rat liver BRL cells.
  • Selectively affects splenic T cells in Wistar rats.