Learn More
Medchemexpress LLC HY-101478 1mg Medchemexpress, Fenobam CAS:57653-26-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1014781MG
Encompass_Preferred
Medchemexpress, HY-101478 1mg Fenobam CAS:57653-26-6 Fenobam is a selective, orally active, and non-competitive mGluR5 antagonist acting at an allosteric modulatory site ( K d values are 54 and 31 nM for rat and human recombinant mGlu5 receptors, respectively). Fenobam displays inverse agonist activity which blocks the mGlu5 receptor basal activity with an IC 50 of 84 nM. Fenobam exerts anxiolytic activity [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2144