Learn More
Enzo Life Sciences Fenobam (5mg). CAS: 57653-26-6
SDP
Supplier:  Enzo Life Sciences ALX550413M005
Encompass_Preferred
Potent and selective non-competitive mGluR5 antagonist that displays inverse agonist properties; blocks mGluR5 constitutive activity in vitro (IC50=87nM). Acts at an allosteric modulatory site shared with MPEP (ALX-550-368) and binds mGluR5 with Kd values of 54nM and 31nM for rat and human receptors respectively. Displays anxiolytic activity following oral administration in vivo. Purity: ≥98% (1H-NMR). Solubility: Soluble in DMSO or methanol. Long Term Storage: +4°C.
Catalog No. 50-200-9556