Learn More
Cayman Chemical Fenobam (hydrate), Purity: ≥98%, CAS: 63540-28-3, 10 mg
SDP
Catalog No. 502989039
Encompass_Preferred
A noncompetitive antagonist and inverse agonist of mGluR5 that binds to an allosteric site; inhibits quisqualate-induced intracellular calcium mobilization in HEK293 cells expressing the human receptor (IC50 = 58 nM); inhibits basal activity of mGluR5 in a cell-based assay (IC50 = 84 nM); reduces stress-induced hyperthermia in mice at 10 and 30 mg/kg; increases drinking time in the Vogel conflict test in rats at 30 mg/kg; induces analgesia in the formalin test in mice at 30 mg/kg
Catalog No. 50-298-9039 Supplier Cayman Chemical Supplier No. 2975910 MG
A noncompetitive antagonist and inverse agonist of mGluR5 that binds to an allosteric site; inhibits quisqualate-induced intracellular calcium mobilization in HEK293 cells expressing the human receptor (IC50 = 58 nM); inhibits basal activity of mGluR5 in a cell-based assay (IC50 = 84 nM); reduces stress-induced hyperthermia in mice at 10 and 30 mg/kg; increases drinking time in the Vogel conflict test in rats at 30 mg/kg; induces analgesia in the formalin test in mice at 30 mg/kg