Learn More
Medchemexpress LLC HY-B0835 250mg Medchemexpress, Fenobucarb CAS:3766-81-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0835250MG
Encompass_Preferred
Medchemexpress, HY-B0835 250mg Fenobucarb CAS:3766-81-2 Fenobucarb is a carbamate insecticide. Fenobucarb induces zebrafish developmental neurotoxicity through pathways involved in inflammation, oxidative stress, degeneration and apoptosis. Fenobucarb is a possible risk factor to cardiovascular and cerebrovascular systems in animals. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2662