Learn More
Medchemexpress LLC HY-17356 200mg Medchemexpress, Fenofibrate CAS:49562-28-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY17356200MG
Encompass_Preferred
Medchemexpress, HY-17356 200mg Fenofibrate CAS:49562-28-9 Fenofibrate is a selective PPARα agonist with an EC50 of 30 μM. Fenofibrate also inhibits human cytochrome P450 isoforms, with IC50s of 0.2, 0.7, 9.7, 4.8 and 142.1 μM for CYP2C19, CYP2B6, CYP2C9, CYP2C8, and CYP3A4, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0761