Learn More
Medchemexpress LLC Fenofibrate | 49562-28-9 | 99.92% | 360.83 | 100 MG
SDP
Catalog No. 5000384698
Encompass_Preferred
Fenofibrate (Standard) is the analytical standard of Fenofibrate, intended for research and analytical applications. It is a selective PPARα agonist with an EC50 of 30 μM and also inhibits human cytochrome P450 isoforms. This analytical standard is commonly used in qualitative, quantitative, and methodological research experiments.
  • Used in HPLC, GC, and MS experiments
  • Reference standard for assay
  • Selective PPARα agonist
  • Inhibits human cytochrome P450 isoforms
Catalog No. 50-003-84698 Supplier Medchemexpress LLC Supplier No. HY17356R100MG
Fenofibrate (Standard) is the analytical standard of Fenofibrate, intended for research and analytical applications. It is a selective PPARα agonist with an EC50 of 30 μM and also inhibits human cytochrome P450 isoforms. This analytical standard is commonly used in qualitative, quantitative, and methodological research experiments.
  • Used in HPLC, GC, and MS experiments
  • Reference standard for assay
  • Selective PPARα agonist
  • Inhibits human cytochrome P450 isoforms