Learn More
Apexbio Technology LLC Fenofibrate, 5g Cas# 49562-28-9
SDP
Supplier:  Apexbio Technology LLC B19435000
Encompass_Preferred
Fenofibrate is PPAR agonist with EC50 of 18 M and 30 M for mouse and human PPAR, respectively. Other sizes are available. Please inqury us for quote.
Catalog No. 50-199-8558