Learn More
Medchemexpress LLC Fenofibrate-d6 | 1092484-56-4 | 99.7% | 366.87 | 1 MG
SDP
Catalog No. 5000385993
Encompass_Preferred
Fenofibrate-d6 is the deuterium labeled version of Fenofibrate. Fenofibrate is recognized as a selective PPARα agonist with an EC50 of 30 μM. It also acts as an inhibitor of various human cytochrome P450 isoforms, demonstrating IC50s of 0.2, 0.7, 9.7, 4.8, and 142.1 μM for CYP2C19, CYP2B6, CYP2C9, CYP2C8, and CYP3A4, respectively.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated, which can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-85993 Supplier Medchemexpress LLC Supplier No. HY17356S1MG
Fenofibrate-d6 is the deuterium labeled version of Fenofibrate. Fenofibrate is recognized as a selective PPARα agonist with an EC50 of 30 μM. It also acts as an inhibitor of various human cytochrome P450 isoforms, demonstrating IC50s of 0.2, 0.7, 9.7, 4.8, and 142.1 μM for CYP2C19, CYP2B6, CYP2C9, CYP2C8, and CYP3A4, respectively.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated, which can affect the pharmacokinetic and metabolic profiles of drugs.