Learn More
Selleck Chemical LLC Fenofibrate NSC-281319-50mg
SDP
Catalog No. 502389587
Encompass_Preferred
Fenofibrate (NSC-281319) is a compound of the fibrate class and fibric acid derivative. Fenofibrate is a selective agonist of PPARα with EC50 of 30 μM. Fenofibrate binds to and inhibits cytochrome P450 epoxygenase (CYP)2C with IC50 of 0.2 μM, 0.7 μM and 9.7 μM for CYP2C19, CYP2B6 and CYP2C9, respectively. Fenofibrate induces autophagy.
Catalog No. 50-238-9587 Supplier Selleck Chemical LLC Supplier No. S179450MG
Fenofibrate (NSC-281319) is a compound of the fibrate class and fibric acid derivative. Fenofibrate is a selective agonist of PPARα with EC50 of 30 μM. Fenofibrate binds to and inhibits cytochrome P450 epoxygenase (CYP)2C with IC50 of 0.2 μM, 0.7 μM and 9.7 μM for CYP2C19, CYP2B6 and CYP2C9, respectively. Fenofibrate induces autophagy.