Learn More
Medchemexpress LLC Fenofibrate (Standard) | 49562-28-9 | 99.9% | 360.83 | 25 MG
SDP
Catalog No. 5000383973
Encompass_Preferred
Fenofibrate (Standard) is the analytical standard of Fenofibrate, intended for research and analytical applications. It is a selective PPARα agonist with an EC50 of 30 μM. It also inhibits human cytochrome P450 isoforms, with IC50s of 0.2, 0.7, 9.7, 4.8 and 142.1 μM for CYP2C19, CYP2B6, CYP2C9, CYP2C8, and CYP3A4, respectively. This analytical standard is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
  • Analytical standard of fenofibrate.
  • Intended for research and analytical applications.
  • Selective PPARα agonist.
  • Inhibits human cytochrome P450 isoforms.
  • Used in qualitative research experiments in HPLC, GC and MS.
  • Used in quantitative research experiments in HPLC, GC and MS.
  • Used in methodological research experiments in HPLC, GC and MS.
Catalog No. 50-003-83973 Supplier Medchemexpress LLC Supplier No. 25MGHY17356R
Fenofibrate (Standard) is the analytical standard of Fenofibrate, intended for research and analytical applications. It is a selective PPARα agonist with an EC50 of 30 μM. It also inhibits human cytochrome P450 isoforms, with IC50s of 0.2, 0.7, 9.7, 4.8 and 142.1 μM for CYP2C19, CYP2B6, CYP2C9, CYP2C8, and CYP3A4, respectively. This analytical standard is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
  • Analytical standard of fenofibrate.
  • Intended for research and analytical applications.
  • Selective PPARα agonist.
  • Inhibits human cytochrome P450 isoforms.
  • Used in qualitative research experiments in HPLC, GC and MS.
  • Used in quantitative research experiments in HPLC, GC and MS.
  • Used in methodological research experiments in HPLC, GC and MS.