Learn More
Apexbio Technology LLC Fenofibric acid 42017-89-0 1g
SDP
Catalog No. 502240518
Encompass_Preferred
Fenofibric acid (CAS 42017-89-0) is a small-molecule agonist targeting peroxisome proliferator-activated receptor alpha (PPAR ) It is designed to modulate lipid metabolism by activating PPAR thereby regulating pathways involved in lipid homeostasis Fenofibric acid exerts its biological activity primarily through PPAR activation It demonstrates agonistic activity with EC50 values of 22 4 M 1 47 M and 1 06 M for different PPAR -dependent responses Additionally fenofibric acid inhibits a specific target with an IC50 value of 48 nM Based on these pharmacological properties fenofibric acid holds research potential in studies of lipid regulation dyslipidemia hypertriglyceridemia and related therapeutic interventions
This item is not returnable. View return policy
Catalog No. 50-224-0518 Supplier Apexbio Technology LLC Supplier No. B61281000
Fenofibric acid (CAS 42017-89-0) is a small-molecule agonist targeting peroxisome proliferator-activated receptor alpha (PPAR ) It is designed to modulate lipid metabolism by activating PPAR thereby regulating pathways involved in lipid homeostasis Fenofibric acid exerts its biological activity primarily through PPAR activation It demonstrates agonistic activity with EC50 values of 22 4 M 1 47 M and 1 06 M for different PPAR -dependent responses Additionally fenofibric acid inhibits a specific target with an IC50 value of 48 nM Based on these pharmacological properties fenofibric acid holds research potential in studies of lipid regulation dyslipidemia hypertriglyceridemia and related therapeutic interventions