Learn More
Medchemexpress LLC Fenofibric acid | 42017-89-0 | 250 MG
SDP
Catalog No. 5000406141
Encompass_Preferred
Fenofibric acid, an active metabolite of fenofibrate, is a PPAR activator. It has EC50s of 22.4 μM for PPARα, 1.47 μM for PPARγ, and 1.06 μM for PPARδ. Fenofibric acid also inhibits COX-2 enzyme activity with an IC50 of 48 nM.
  • PPAR activator
  • Inhibits COX-2 enzyme activity
Catalog No. 50-004-06141 Supplier Medchemexpress LLC Supplier No. 250MGHYB0760R
Fenofibric acid, an active metabolite of fenofibrate, is a PPAR activator. It has EC50s of 22.4 μM for PPARα, 1.47 μM for PPARγ, and 1.06 μM for PPARδ. Fenofibric acid also inhibits COX-2 enzyme activity with an IC50 of 48 nM.
  • PPAR activator
  • Inhibits COX-2 enzyme activity