Learn More
Medchemexpress LLC Fenofibric acid | 42017-89-0 | 99.9% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0760500MG
Encompass_Preferred
Fenofibric acid is an active metabolite of fenofibrate and acts as a PPAR activator. It has EC50 values of 22.4 μM for PPARα, 1.47 μM for PPARγ, and 1.06 μM for PPARδ. It also inhibits COX-2 enzyme activity with an IC50 of 48 nM. This product is for research use only.
  • Inhibits COX-2 enzyme dose-dependently with an IC50 of 48 nM.
  • Reduces abundance of AOX1 protein in HepG2 cells at 500 nM.
  • Decreases JNK1/2, c-Jun, and p38 MAPK phosphorylation at 100 μM.
  • Prevents reactive oxygen species accumulation, ER stress, and blood retinal barrier disruption.
  • Activates IGF-IR/Akt/ERK1/2-mediated survival signaling pathways.
  • Demonstrates anti-inflammatory activity in Wistar rats at doses of 1, 5, 10 mg/kg (p.o.).
Catalog No. 50-004-08167