Learn More
Medchemexpress LLC Fenofibric acid | 42017-89-0 | 99.9% | 500 MG
SDP
Catalog No. 5000408937
Encompass_Preferred
Fenofibric acid is an active metabolite of fenofibrate and a PPAR activator, showing EC50s of 22.4 μM for PPARα, 1.47 μM for PPARγ, and 1.06 μM for PPARδ. It also inhibits COX-2 enzyme activity with an IC50 of 48 nM. This product is for research use only.
  • Potently activates PPARα, PPARγ, and PPARδ
  • Inhibits COX-2 enzyme activity
  • Shows anti-inflammatory activity in Wistar rats
  • Reduces abundance of AOX1 protein in HepG2 cells
  • Decreases JNK1/2, c-Jun, and p38 MAPK phosphorylation
  • Prevents accumulation of reactive oxygen species, ER stress, and disruption of the blood retinal barrier
  • Activates IGF-IR/Akt/ERK1/2-mediated survival signaling pathways
  • High purity of 99.94%
  • Available in various forms: solid and solution in DMSO
Catalog No. 50-004-08937 Supplier Medchemexpress LLC Supplier No. HYB0760R500MG
Fenofibric acid is an active metabolite of fenofibrate and a PPAR activator, showing EC50s of 22.4 μM for PPARα, 1.47 μM for PPARγ, and 1.06 μM for PPARδ. It also inhibits COX-2 enzyme activity with an IC50 of 48 nM. This product is for research use only.
  • Potently activates PPARα, PPARγ, and PPARδ
  • Inhibits COX-2 enzyme activity
  • Shows anti-inflammatory activity in Wistar rats
  • Reduces abundance of AOX1 protein in HepG2 cells
  • Decreases JNK1/2, c-Jun, and p38 MAPK phosphorylation
  • Prevents accumulation of reactive oxygen species, ER stress, and disruption of the blood retinal barrier
  • Activates IGF-IR/Akt/ERK1/2-mediated survival signaling pathways
  • High purity of 99.94%
  • Available in various forms: solid and solution in DMSO