Learn More
Medchemexpress LLC Fenofibric acid | 42017-89-0 | MFCD00792461 | 99.57% | 100 MG
SDP
Catalog No. 5000407071
Encompass_Preferred
Fenofibric acid (Standard) is the analytical standard of Fenofibric acid. This product is intended for research and analytical applications. Fenofibric acid, an active metabolite of fenofibrate, is a PPARα, PPARγ, and PPARδ activator with EC50s of 22.4 μM, 1.47 μM, and 1.06 μM, respectively. It also inhibits COX-2 enzyme activity with an IC50 of 48 nM.
  • Intended for research and analytical applications
  • Active metabolite of fenofibrate
  • Acts as a PPARα, PPARγ, and PPARδ activator
  • Inhibits COX-2 enzyme activity
Catalog No. 50-004-07071 Supplier Medchemexpress LLC Supplier No. HYB0760R100MG
Fenofibric acid (Standard) is the analytical standard of Fenofibric acid. This product is intended for research and analytical applications. Fenofibric acid, an active metabolite of fenofibrate, is a PPARα, PPARγ, and PPARδ activator with EC50s of 22.4 μM, 1.47 μM, and 1.06 μM, respectively. It also inhibits COX-2 enzyme activity with an IC50 of 48 nM.
  • Intended for research and analytical applications
  • Active metabolite of fenofibrate
  • Acts as a PPARα, PPARγ, and PPARδ activator
  • Inhibits COX-2 enzyme activity