Learn More
Medchemexpress LLC Fenofibric acid-d6 | 1092484-69-9 | 99.6% | 5 MG
SDP
Catalog No. 5000405508
Encompass_Preferred
Fenofibric acid-d6 (FNF acid-d6) is the deuterium-labeled form of Fenofibric acid. Fenofibric acid, an active metabolite of fenofibrate, acts as a PPAR activator with EC50s of 22.4 μM for PPARα, 1.47 μM for PPARγ, and 1.06 μM for PPARδ. It also inhibits COX-2 enzyme activity with an IC50 of 48 nM. Deuterium substitution in drug molecules is of interest due to its potential impact on pharmacokinetic and metabolic profiles.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • For research use only.
Catalog No. 50-004-05508 Supplier Medchemexpress LLC Supplier No. HYB0760S5MG
Fenofibric acid-d6 (FNF acid-d6) is the deuterium-labeled form of Fenofibric acid. Fenofibric acid, an active metabolite of fenofibrate, acts as a PPAR activator with EC50s of 22.4 μM for PPARα, 1.47 μM for PPARγ, and 1.06 μM for PPARδ. It also inhibits COX-2 enzyme activity with an IC50 of 48 nM. Deuterium substitution in drug molecules is of interest due to its potential impact on pharmacokinetic and metabolic profiles.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • For research use only.