Learn More
Medchemexpress LLC Fenofibric acid-d6 | 1092484-69-9 | MFCD06658651 | 99.6% | 1 MG
SDP
Catalog No. 5000408154
Encompass_Preferred
Fenofibric acid-d6 is the deuterium-labeled form of Fenofibric acid, which is an active metabolite of fenofibrate. It acts as a PPAR activator and also inhibits COX-2 enzyme activity. This compound is useful as a tracer or as an internal standard for quantitative analysis.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Active metabolite of fenofibrate
  • PPAR activator
  • Inhibits COX-2 enzyme activity
Catalog No. 50-004-08154 Supplier Medchemexpress LLC Supplier No. HYB0760S1MG
Fenofibric acid-d6 is the deuterium-labeled form of Fenofibric acid, which is an active metabolite of fenofibrate. It acts as a PPAR activator and also inhibits COX-2 enzyme activity. This compound is useful as a tracer or as an internal standard for quantitative analysis.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Active metabolite of fenofibrate
  • PPAR activator
  • Inhibits COX-2 enzyme activity