Learn More
Lkt Laboratories Fenoldopam Mesylate, ≥98%, 500MG
SDP
Supplier:  Lkt Laboratories F1654500MG
Encompass_Preferred
Benzazepine derivative; D1 partial agonist, potential α1-adrenergic antagonist. CAS: 67227-57-0. Additional sizes available upon request. Ships from Saint Paul, MN.
Catalog No. 50-153-4918