Learn More
Apexbio Technology LLC Fenoldopam (mesylate), 50mg CAS# 67227-57-0
SDP
Supplier:  Apexbio Technology LLC B868150
Encompass_Preferred
Fenoldopam mesylate is a selective dopamine D1 receptor agonist used as an antihypertensive drug. Fenoldopam mesylate causes peripheral vasodilation by stimulating dopamine-1 receptor and 2-adrenoceptor, increasing renal blood flow. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8462