Learn More
Medchemexpress LLC Fenoldopam mesylate | 67227-57-0 | 99.94% | 5 MG
SDP
Catalog No. 5000407086
Encompass_Preferred
Fenoldopam mesylate (SKF-82526) is a D1 receptor agonist and a novel lysine-specific demethylase 1 (LSD1) inhibitor. It exhibits anti-hypertensive effects, anti-cancer cell proliferation activity, and can induce cell apoptosis. This compound is suitable for research use only.
  • Functions as a D1 receptor agonist
  • Acts as a novel LSD1 inhibitor with an IC50 of 0.8974 μM
  • Exhibits anti-hypertensive effects
  • Demonstrates anti-cancer cell proliferation activity
  • Can induce cell apoptosis
  • Shows antiproliferative activity against RCC cell lines
  • Induces cell apoptosis in ACHN cells in a dose-dependent manner
Catalog No. 50-004-07086 Supplier Medchemexpress LLC Supplier No. HYB0735A5MG
Fenoldopam mesylate (SKF-82526) is a D1 receptor agonist and a novel lysine-specific demethylase 1 (LSD1) inhibitor. It exhibits anti-hypertensive effects, anti-cancer cell proliferation activity, and can induce cell apoptosis. This compound is suitable for research use only.
  • Functions as a D1 receptor agonist
  • Acts as a novel LSD1 inhibitor with an IC50 of 0.8974 μM
  • Exhibits anti-hypertensive effects
  • Demonstrates anti-cancer cell proliferation activity
  • Can induce cell apoptosis
  • Shows antiproliferative activity against RCC cell lines
  • Induces cell apoptosis in ACHN cells in a dose-dependent manner