Learn More
Selleck Chemical LLC Fenoldopam mesylate S4618-200mg
SDP
Catalog No. 503111464
Encompass_Preferred
Fenoldopam (SKF 82526 Corlopam) is a selective dopamine-1 receptor (DA1) agonist with natriuretic/diuretic properties It lowers blood pressure through arteriolar vasodilation
Catalog No. 50-311-1464 Supplier Selleck Chemical LLC Supplier No. S4618200MG
Fenoldopam (SKF 82526 Corlopam) is a selective dopamine-1 receptor (DA1) agonist with natriuretic/diuretic properties It lowers blood pressure through arteriolar vasodilation