Learn More
Cayman Chemical Fenoldopam (mesylate), Purity: ≥95%, CAS: 67227-57-0, 50 mg
SDP
Catalog No. 502967699
Encompass_Preferred
An agonist of dopamine D1A (D1R) and D1B (D5R) receptors (Kds = 17 and 11 nM, respectively); alters hemodynamic properties, including hypertension, in animals.
Catalog No. 50-296-7699 Supplier Cayman Chemical Supplier No. 1762950 MG
An agonist of dopamine D1A (D1R) and D1B (D5R) receptors (Kds = 17 and 11 nM, respectively); alters hemodynamic properties, including hypertension, in animals.