Learn More
Apexbio Technology LLC Fenoprofen Calcium 34597-40-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062350
Encompass_Preferred
Fenoprofen Calcium is a non-steroidal anti-inflammatory drug (NSAID) belonging to the phenylpropionic acid class Its mechanism of action involves inhibiting the cyclooxygenase (COX) enzymes resulting in decreased synthesis of prostaglandins inflammatory mediators centrally associated with pain inflammation and fever Due to its COX-mediated pharmacological profile Fenoprofen Calcium is frequently utilized in biomedical research particularly within inflammation-driven disease models including arthritis Researchers routinely employ this compound to investigate the pathways involved in inflammatory signaling prostaglandin biosynthesis and related pain mechanisms providing insights into inflammatory responses and analgesic cellular processes
Catalog No. 50-000-62350 Supplier Apexbio Technology LLC Supplier No. B19445.1
Fenoprofen Calcium is a non-steroidal anti-inflammatory drug (NSAID) belonging to the phenylpropionic acid class Its mechanism of action involves inhibiting the cyclooxygenase (COX) enzymes resulting in decreased synthesis of prostaglandins inflammatory mediators centrally associated with pain inflammation and fever Due to its COX-mediated pharmacological profile Fenoprofen Calcium is frequently utilized in biomedical research particularly within inflammation-driven disease models including arthritis Researchers routinely employ this compound to investigate the pathways involved in inflammatory signaling prostaglandin biosynthesis and related pain mechanisms providing insights into inflammatory responses and analgesic cellular processes