Learn More
Medchemexpress LLC Fenoprofen Calcium hydrate | 71720-56-4 | MFCD00242722 | 99.52% | 1 ML
SDP
Catalog No. 5000407024
Encompass_Preferred
Fenoprofen Calcium hydrate is a nonsteroidal anti-inflammatory agent (NSAID) that inhibits cyclooxygenase (COX). It also acts as a positive allosteric modulator (PAM) of melanocortin receptors (MCRs) and increases ERK1/2 activation in HEK293T cells. Fenoprofen Calcium hydrate exhibits anti-arthritic activities, making it suitable for studies on rheumatoid arthritis and osteoarthritis.
  • Inhibits cyclooxygenase (COX)
  • Positive allosteric modulator of melanocortin receptors (MCRs)
  • Increases ERK1/2 activation in HEK293T cells
  • Exhibits anti-arthritic activities
  • Suitable for studies on rheumatoid arthritis and osteoarthritis
Catalog No. 50-004-07024 Supplier Medchemexpress LLC Supplier No. HYB0288B10MM/1ML
Fenoprofen Calcium hydrate is a nonsteroidal anti-inflammatory agent (NSAID) that inhibits cyclooxygenase (COX). It also acts as a positive allosteric modulator (PAM) of melanocortin receptors (MCRs) and increases ERK1/2 activation in HEK293T cells. Fenoprofen Calcium hydrate exhibits anti-arthritic activities, making it suitable for studies on rheumatoid arthritis and osteoarthritis.
  • Inhibits cyclooxygenase (COX)
  • Positive allosteric modulator of melanocortin receptors (MCRs)
  • Increases ERK1/2 activation in HEK293T cells
  • Exhibits anti-arthritic activities
  • Suitable for studies on rheumatoid arthritis and osteoarthritis