Learn More
TARGETMOL CHEMICALS INC Fenoprofen Calcium hydrate
SDP
Catalog No. 502880646
Encompass_Preferred
Also available in 1 g, 1 mL, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Fenoprofen calcium is a propionic acid derivative that is used as a non-steroidal anti-inflammatory agent. Purity 99.29%
Catalog No. 50-288-0646 Supplier TARGETMOL CHEMICALS INC Supplier No. T0184500MG
Also available in 1 g, 1 mL, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Fenoprofen calcium is a propionic acid derivative that is used as a non-steroidal anti-inflammatory agent. Purity 99.29%