Learn More
Cayman Chemical Fenoprofen (calcium salt hydrate), Purity: ≥98%, CAS: 71720-56-4, 25 g
SDP
Catalog No. 502994813
Encompass_Preferred
An NSAID; inhibits spontaneous and estradiol-stimulated PGF2α release in isolated rat uterine horns; reduces serum PGF2α levels in rats (10 mg/kg); reduces acetic acid-induced writhing and stretching as well as formalin-induced licking behaviors, indicating analgesic activity in mice; induces formation of gastric lesions in mice
Catalog No. 50-299-4813 Supplier Cayman Chemical Supplier No. 2383525 G
An NSAID; inhibits spontaneous and estradiol-stimulated PGF2α release in isolated rat uterine horns; reduces serum PGF2α levels in rats (10 mg/kg); reduces acetic acid-induced writhing and stretching as well as formalin-induced licking behaviors, indicating analgesic activity in mice; induces formation of gastric lesions in mice