Learn More
Medchemexpress LLC Fenoterol-d6 (hydrobromide) | 1286129-04-1 | 98.0% | 1 MG
SDP
Catalog No. 5000408396
Encompass_Preferred
Fenoterol-d6 hydrobromide is the deuterium-labeled version of Fenoterol hydrobromide, a sympathomimetic agent and selective β2-adrenoceptor agonist. It functions as an effective bronchodilator in research contexts, particularly for bronchospasm linked to asthma, bronchitis, and other obstructive airway diseases. This compound is also suitable for use as a tracer and internal standard in quantitative analysis methods such as NMR, GC-MS, or LC-MS.
  • Deuterium-labeled version of Fenoterol hydrobromide
  • Acts as a selective and orally active β2-adrenoceptor agonist
  • Effective bronchodilator for research on bronchospasm associated with respiratory diseases
  • Can be used as a tracer
  • Suitable as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
  • Deuteration may influence pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-08396 Supplier Medchemexpress LLC Supplier No. HYB0976AS1MG
Fenoterol-d6 hydrobromide is the deuterium-labeled version of Fenoterol hydrobromide, a sympathomimetic agent and selective β2-adrenoceptor agonist. It functions as an effective bronchodilator in research contexts, particularly for bronchospasm linked to asthma, bronchitis, and other obstructive airway diseases. This compound is also suitable for use as a tracer and internal standard in quantitative analysis methods such as NMR, GC-MS, or LC-MS.
  • Deuterium-labeled version of Fenoterol hydrobromide
  • Acts as a selective and orally active β2-adrenoceptor agonist
  • Effective bronchodilator for research on bronchospasm associated with respiratory diseases
  • Can be used as a tracer
  • Suitable as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
  • Deuteration may influence pharmacokinetic and metabolic profiles of drugs