Learn More
Medchemexpress LLC Fenoterol hydrobromide | 1944-12-3 | 100.0% | 1 ML
SDP
Catalog No. 5000408395
Encompass_Preferred
Fenoterol hydrobromide (Th-1165a) is a sympathomimetic agent that acts as a selective and orally active β2-adrenoceptor agonist. It is an effective bronchodilator, making it suitable for research into bronchospasm associated with asthma, bronchitis, and other obstructive airway diseases.
  • Selective and orally active β2-adrenoceptor agonist
  • Functions as an effective bronchodilator
  • Used in research for bronchospasm associated with asthma, bronchitis, and other obstructive airway diseases
Catalog No. 50-004-08395 Supplier Medchemexpress LLC Supplier No. HYB0976A10MM/1ML
Fenoterol hydrobromide (Th-1165a) is a sympathomimetic agent that acts as a selective and orally active β2-adrenoceptor agonist. It is an effective bronchodilator, making it suitable for research into bronchospasm associated with asthma, bronchitis, and other obstructive airway diseases.
  • Selective and orally active β2-adrenoceptor agonist
  • Functions as an effective bronchodilator
  • Used in research for bronchospasm associated with asthma, bronchitis, and other obstructive airway diseases