Learn More
Cayman Chemical Fenoterol (hydrobromide), Purity: ≥98%, CAS: 1944-12-3, 10 mg
SDP
Catalog No. 502985565
Encompass_Preferred
An analytical reference standard categorized as a β2-adrenergic receptor agonist; has been used as a performance-enhancing drug in sports doping and is banned by WADA; intended for research and forensic applications
Catalog No. 50-298-5565 Supplier Cayman Chemical Supplier No. 2129310 MG
An analytical reference standard categorized as a β2-adrenergic receptor agonist; has been used as a performance-enhancing drug in sports doping and is banned by WADA; intended for research and forensic applications