Learn More
Medchemexpress LLC Fenoverine-d8 | 00-00-0 | MFCD00865242 | 98.9% | 467.61 g·mol⁻1 | C26H17D8N3O3S | 10 MG
SDP
Catalog No. 5000240603
Encompass_Preferred
Fenoverine-d8 is a deuterium-labeled analytical standard of fenoverine used as a stable isotope internal standard in quantitative LC-MS and other analytical assays. It is provided with high purity and defined molecular properties for method development, validation, and pharmacokinetic or bioanalysis studies.
  • Deuterium-labeled internal standard for LC-MS quantitation.
  • High purity suitable for analytical applications.
  • Characterized molecular weight and formula for accurate identification.
  • Available in multiple small package sizes for method development.
  • Provides improved accuracy in pharmacokinetic and bioanalysis studies.
Catalog No. 50-002-40603 Supplier Medchemexpress LLC Supplier No. HY107349S10MG
Fenoverine-d8 is a deuterium-labeled analytical standard of fenoverine used as a stable isotope internal standard in quantitative LC-MS and other analytical assays. It is provided with high purity and defined molecular properties for method development, validation, and pharmacokinetic or bioanalysis studies.
  • Deuterium-labeled internal standard for LC-MS quantitation.
  • High purity suitable for analytical applications.
  • Characterized molecular weight and formula for accurate identification.
  • Available in multiple small package sizes for method development.
  • Provides improved accuracy in pharmacokinetic and bioanalysis studies.