Learn More
Medchemexpress LLC Fenoverine-d8 1mg | 1MG
SDP
Catalog No. 5000240976
Encompass_Preferred
Fenoverine-d8 is the deuterium labeled Fenoverine Fenoverine (Spasmopriv) is an orally active and powerful anti-spasmodic drug Fenoverine reduces calcium ion influx by suppressing voltage dependent calcium channels Fenoverine has antiemetic analgesic and antidiarrheal effects Fenoverine induces rhabdomyolysis Fenoverine can be used to study smooth muscle overexcitation related conditions such as gastrointestinal spasms[1 [2 [3
Catalog No. 50-002-40976 Supplier Medchemexpress LLC Supplier No. HY107349S1MG
Fenoverine-d8 is the deuterium labeled Fenoverine Fenoverine (Spasmopriv) is an orally active and powerful anti-spasmodic drug Fenoverine reduces calcium ion influx by suppressing voltage dependent calcium channels Fenoverine has antiemetic analgesic and antidiarrheal effects Fenoverine induces rhabdomyolysis Fenoverine can be used to study smooth muscle overexcitation related conditions such as gastrointestinal spasms[1 [2 [3