Learn More
Medchemexpress LLC Fenoxaprop-P-ethyl | 71283-80-2 | MFCD01310637 | 99.6% | 5 G
SDP
Catalog No. 5000411237
Encompass_Preferred
Fenoxaprop-P-ethyl is a phenoxy propionic acid herbicide with a heterocyclic epoxide structure. This product presents as a solid and is stable under recommended storage conditions, exhibiting a melting/freezing point between 79-84°C.
  • Functions as a herbicide
  • Acts on the metabolic enzyme/protease pathway
  • Solid appearance
  • Melting/freezing point between 79-84°C
  • Stable under recommended storage conditions
Catalog No. 50-004-11237 Supplier Medchemexpress LLC Supplier No. HYB20135G
Fenoxaprop-P-ethyl is a phenoxy propionic acid herbicide with a heterocyclic epoxide structure. This product presents as a solid and is stable under recommended storage conditions, exhibiting a melting/freezing point between 79-84°C.
  • Functions as a herbicide
  • Acts on the metabolic enzyme/protease pathway
  • Solid appearance
  • Melting/freezing point between 79-84°C
  • Stable under recommended storage conditions