Learn More
Medchemexpress LLC Fenoxaprop-P-ethyl (Standard) | 71283-80-2 | 99.5% | 25 MG
SDP
Catalog No. 5000411256
Encompass_Preferred
Fenoxaprop-P-ethyl (Standard) is the analytical standard for Fenoxaprop-P-ethyl, intended for research and analytical applications. It is commonly used as a reference standard in qualitative, quantitative, and methodological research experiments involving HPLC, GC, and MS.
  • Analytical standard for Fenoxaprop-P-ethyl
  • Suitable for research and analytical applications
  • Used as a reference standard
  • Commonly used in HPLC experiments
  • Commonly used in GC experiments
  • Commonly used in MS experiments
Catalog No. 50-004-11256 Supplier Medchemexpress LLC Supplier No. 25MGHYB2013R
Fenoxaprop-P-ethyl (Standard) is the analytical standard for Fenoxaprop-P-ethyl, intended for research and analytical applications. It is commonly used as a reference standard in qualitative, quantitative, and methodological research experiments involving HPLC, GC, and MS.
  • Analytical standard for Fenoxaprop-P-ethyl
  • Suitable for research and analytical applications
  • Used as a reference standard
  • Commonly used in HPLC experiments
  • Commonly used in GC experiments
  • Commonly used in MS experiments