Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fenoxaprop-P-ethyl (Standard) | 71283-80-2 | MFCD01310637 | 99.6% | 10 G
SDP

Catalog No. 5000412176
Encompass_Preferred

Fenoxaprop-P-ethyl is a phenoxy propionic acid herbicide with a heterocyclic epoxide structure. It is intended for research use only and has not been fully validated for medical applications.

  • Targeted as a herbicide
  • Acts on the metabolic enzyme/protease pathway
  • Molecular weight: 361.78
  • Molecular formula: C18H16ClNO5
  • Appearance: Solid
  • Color: White to off-white
  • Purity: 99.55%
  • Storage for powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 2 years, -20°C for 1 year

Catalog No. 50-004-12176 Supplier Medchemexpress LLC Supplier No. HYB201310G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fenoxaprop-P-ethyl is a phenoxy propionic acid herbicide with a heterocyclic epoxide structure. It is intended for research use only and has not been fully validated for medical applications.

  • Targeted as a herbicide
  • Acts on the metabolic enzyme/protease pathway
  • Molecular weight: 361.78
  • Molecular formula: C18H16ClNO5
  • Appearance: Solid
  • Color: White to off-white
  • Purity: 99.55%
  • Storage for powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 2 years, -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.