Learn More
Cayman Chemical Fenoxycarb-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 2468639-22-5, 1 mg
SDP
Catalog No. 502915895
Encompass_Preferred
An internal standard for the quantification of fenoxycarb by GC- or LC-MS
Catalog No. 50-291-5895 Supplier Cayman Chemical Supplier No. 395981 MG
An internal standard for the quantification of fenoxycarb by GC- or LC-MS