Learn More
Cayman Chemical Fenpropathrin, Purity: ≥95%, CAS: 39515-41-8, 50 mg
SDP
Catalog No. 502948127
Encompass_Preferred
A pyrethroid insecticide; a slow activator of Nav1.8; decreases the number of Colorado potato beetle (L. decemlineata) adults and third stage larvae at 0.01%; has been found in watershed sediment samples
Catalog No. 50-294-8127 Supplier Cayman Chemical Supplier No. 3447850 MG
A pyrethroid insecticide; a slow activator of Nav1.8; decreases the number of Colorado potato beetle (L. decemlineata) adults and third stage larvae at 0.01%; has been found in watershed sediment samples