Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fenpropathrin-d5 | 00-00-0 | 98.0% | 354.45 g/mol | C22H18D5NO3 | 5 MG
SDP

Catalog No. 5000263009
Encompass_Preferred

Fenpropathrin-d5 is the deuterium-labeled (d5) analogue of the synthetic pyrethroid insecticide fenpropathrin, provided as an analytical isotope-labeled standard for use as an internal standard, tracer, or reference in quantitative workflows including LC-MS/MS. The material is supplied at high purity and includes manufacturer guidance for solubility and storage to support reproducible analytical results.

  • Deuterium-labeled standard for accurate quantitation.
  • High purity (98.0%) for analytical reliability.
  • Suitable for LC-MS/MS internal standard or tracer studies.
  • Soluble in DMSO at 100 mg/mL; use freshly opened DMSO and ultrasonic agitation.
  • Store at 4°C protected from light; in solution store at -80°C up to 6 months.

Catalog No. 50-002-63009 Supplier Medchemexpress LLC Supplier No. HY123178S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fenpropathrin-d5 is the deuterium-labeled (d5) analogue of the synthetic pyrethroid insecticide fenpropathrin, provided as an analytical isotope-labeled standard for use as an internal standard, tracer, or reference in quantitative workflows including LC-MS/MS. The material is supplied at high purity and includes manufacturer guidance for solubility and storage to support reproducible analytical results.

  • Deuterium-labeled standard for accurate quantitation.
  • High purity (98.0%) for analytical reliability.
  • Suitable for LC-MS/MS internal standard or tracer studies.
  • Soluble in DMSO at 100 mg/mL; use freshly opened DMSO and ultrasonic agitation.
  • Store at 4°C protected from light; in solution store at -80°C up to 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.