Learn More
Medchemexpress LLC Fenpropathrin-d5 | 98.0% | 354.45 | 1 MG
SDP
Catalog No. 5000438413
Encompass_Preferred
Fenpropathrin-d5 is the deuterium labeled Fenpropathrin, a synthetic pyrethroid insecticide used in agriculture. It may progressively induce parkinsonian symptoms. Deuteration, incorporating stable heavy isotopes, is used as a tracer for quantitation during drug development and has the potential to affect pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled Fenpropathrin.
  • Synthetic pyrethroid insecticide.
Catalog No. 50-004-38413 Supplier Medchemexpress LLC Supplier No. HY123178S1MG
Fenpropathrin-d5 is the deuterium labeled Fenpropathrin, a synthetic pyrethroid insecticide used in agriculture. It may progressively induce parkinsonian symptoms. Deuteration, incorporating stable heavy isotopes, is used as a tracer for quantitation during drug development and has the potential to affect pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled Fenpropathrin.
  • Synthetic pyrethroid insecticide.