Learn More
Cayman Chemical Fenpropimorph, Purity: ≥98%, CAS: 67564-91-4, 100 mg
SDP
Catalog No. 502985377
Encompass_Preferred
A fungicide that inhibits the sterol pathway
Catalog No. 50-298-5377 Supplier Cayman Chemical Supplier No. 20875100 MG
A fungicide that inhibits the sterol pathway