Learn More
Medchemexpress LLC Fenpropimorph | 67564-91-4 | 98.7% | 1 MG
SDP
Catalog No. 5000340763
Encompass_Preferred
Fenpropimorph is a fungicide that inhibits the sterol pathway. It inhibits δ8-δ7-sterol isomerase in yeast at low concentrations, and δ14-sterol reductase at higher levels, which prevents ergosterol biosynthesis. Fenpropimorph also inhibits sterol synthesis in certain plants and mammalian cells.
  • Fungicide
  • Inhibits the sterol pathway
  • Prevents ergosterol biosynthesis
  • Inhibits sterol synthesis in certain plants and mammalian cells
Catalog No. 50-003-40763 Supplier Medchemexpress LLC Supplier No. HY1280331MG
Fenpropimorph is a fungicide that inhibits the sterol pathway. It inhibits δ8-δ7-sterol isomerase in yeast at low concentrations, and δ14-sterol reductase at higher levels, which prevents ergosterol biosynthesis. Fenpropimorph also inhibits sterol synthesis in certain plants and mammalian cells.
  • Fungicide
  • Inhibits the sterol pathway
  • Prevents ergosterol biosynthesis
  • Inhibits sterol synthesis in certain plants and mammalian cells