Learn More
Medchemexpress LLC Fenpropimorph | 67564-91-4 | 98.7% | 10 MG
SDP
Catalog No. 5000341684
Encompass_Preferred
Fenpropimorph is a fungicide that operates by inhibiting the sterol pathway. It blocks δ8-δ7-sterol isomerase at lower concentrations and δ14-sterol reductase at higher concentrations in yeast, which prevents ergosterol biosynthesis. Fenpropimorph also has an inhibitory effect on sterol synthesis in certain plants and mammalian cells.
  • Inhibits the sterol pathway
  • Blocks delta8-delta7-sterol isomerase
  • Blocks delta14-sterol reductase
  • Prevents ergosterol biosynthesis
  • Inhibits sterol synthesis in plants
  • Inhibits sterol synthesis in mammalian cells
  • For research use only
Catalog No. 50-003-41684 Supplier Medchemexpress LLC Supplier No. HY12803310MG
Fenpropimorph is a fungicide that operates by inhibiting the sterol pathway. It blocks δ8-δ7-sterol isomerase at lower concentrations and δ14-sterol reductase at higher concentrations in yeast, which prevents ergosterol biosynthesis. Fenpropimorph also has an inhibitory effect on sterol synthesis in certain plants and mammalian cells.
  • Inhibits the sterol pathway
  • Blocks delta8-delta7-sterol isomerase
  • Blocks delta14-sterol reductase
  • Prevents ergosterol biosynthesis
  • Inhibits sterol synthesis in plants
  • Inhibits sterol synthesis in mammalian cells
  • For research use only