Learn More
FUJIFILM Biosciences Fenpyrazamine Standard
Catalog No. 5000450407
Encompass_Preferred
Fenpyrazamine Standard is a high-purity analytical reference material designed for pesticide residue analysis Supplied as a white crystalline powder it offers 98 0% purity as determined by both HPLC and qNMR methods This compound is practically insoluble in water but readily soluble in organic solvents such as acetone and ethanol It is intended for research use only and is especially suitable for environmental and food safety laboratories requiring accurate quantification of fenpyrazamine residues The product must be stored in a tightly sealed container at 2 10C protected from light and moisture Classified as environmentally hazardous UN3077 it should be handled in accordance with all relevant safety and transport regulations
Catalog No. 50-004-50407 Supplier FUJIFILM Biosciences Supplier No. 06906171
Fenpyrazamine Standard is a high-purity analytical reference material designed for pesticide residue analysis Supplied as a white crystalline powder it offers 98 0% purity as determined by both HPLC and qNMR methods This compound is practically insoluble in water but readily soluble in organic solvents such as acetone and ethanol It is intended for research use only and is especially suitable for environmental and food safety laboratories requiring accurate quantification of fenpyrazamine residues The product must be stored in a tightly sealed container at 2 10C protected from light and moisture Classified as environmentally hazardous UN3077 it should be handled in accordance with all relevant safety and transport regulations