Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fenquizone | 20287-37-0 | MFCD00867349 | 99.6% | 337.78 g·mol^-1 | C14H12ClN3O3S | 10MM 1ML
SDP

Catalog No. 5000239263
Encompass_Preferred

Fenquizone is a thiazide-like diuretic research compound offered as a 10 mM solution in 1 mL DMSO or as a solid standard for analytical use. It has chemical formula C14H12ClN3O3S, molecular weight 337.78 g·mol^-1, and CAS number 20287-37-0. The product is intended for research use only and is not for human or veterinary use.

  • Thiazide-like diuretic useful for edema and hypertension research.
  • Available as a 10 mM solution in 1 mL DMSO and as a solid/standard format.
  • High purity (99.6%) suitable for analytical and research applications.
  • Molecular formula C14H12ClN3O3S and molecular weight 337.78 g·mol^-1.
  • Identified by CAS number 20287-37-0 for unambiguous reference.

Catalog No. 50-002-39263 Supplier Medchemexpress LLC Supplier No. HY12617910MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fenquizone is a thiazide-like diuretic research compound offered as a 10 mM solution in 1 mL DMSO or as a solid standard for analytical use. It has chemical formula C14H12ClN3O3S, molecular weight 337.78 g·mol^-1, and CAS number 20287-37-0. The product is intended for research use only and is not for human or veterinary use.

  • Thiazide-like diuretic useful for edema and hypertension research.
  • Available as a 10 mM solution in 1 mL DMSO and as a solid/standard format.
  • High purity (99.6%) suitable for analytical and research applications.
  • Molecular formula C14H12ClN3O3S and molecular weight 337.78 g·mol^-1.
  • Identified by CAS number 20287-37-0 for unambiguous reference.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.