Learn More
Selleck Chemical LLC Fenretinide 25mg 65646-68-6 4-hydroxy(phenyl)retinamide, 4-HPR, MK-4016
SDP
Supplier:  Selleck Chemical LLC S523325MG
Encompass_Preferred
Fenretinide (4-hydroxy(phenyl)retinamide, 4-HPR, MK-4016) is a synthetic derivative of retinoic acid that has a broad-spectrum of cytotoxic activity against primary tumor cells, cell lines, and/or xenografts of various cancers (including neuroblastoma). Fenretinide (4-hydroxy(phenyl)retinamide; 4-HPR) is a synthetic derivative of retinoic acid, fenretinide can bind to retinoic acid receptor (RAR) and retinoid X receptor (RXR) at concentrations necessary to induce cell death. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8280