Learn More
Medchemexpress LLC HY-15373 100mg , Fenretinide 4-HPR;(4-Hydroxyphenyl)retinamide CAS:65646-68-6 Purity:98%
SDP
Catalog No. 501493086
Encompass_Preferred
Medchemexpress, HY-15373 100mg Fenretinide 4-HPR;(4-Hydroxyphenyl)retinamide CAS:65646-68-6 Formula:C26H33NO2 Purity:98% Fenretinide is a synthetic retinoid deriverative, binding to the retinoic acid receptors ( RAR ) at concentrations necessary to induce cell death. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3086 Supplier Medchemexpress LLC Supplier No. HY15373100MG
Medchemexpress, HY-15373 100mg Fenretinide 4-HPR;(4-Hydroxyphenyl)retinamide CAS:65646-68-6 Formula:C26H33NO2 Purity:98% Fenretinide is a synthetic retinoid deriverative, binding to the retinoic acid receptors ( RAR ) at concentrations necessary to induce cell death. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.