Learn More
Cayman Chemical FenretInIde-d4 500ug
SDP
Supplier:  Cayman Chemical 26489500 UG
Encompass_Preferred
An internal standard for the quantification of fenretinide by GC- or LC-MS
Catalog No. 50-293-4362