Learn More
Medchemexpress LLC Fenspiride hydrochloride | 5053-08-7 | 99.1% | C15H21ClN2O2 | 1 ML
SDP
Catalog No. 5000432930
Encompass_Preferred
Fenspiride hydrochloride is an orally active, non-steroidal anti-inflammatory agent that acts as an antagonist of the H1-histamine receptor. It inhibits phosphodiesterase 3 (PDE3), phosphodiesterase 4 (PDE4), and phosphodiesterase 5 (PDE5) activities. This compound can be used for research related to respiratory diseases.
  • Antagonist of the H1-histamine receptor
  • Inhibits phosphodiesterase 3, phosphodiesterase 4, and phosphodiesterase 5 activities
  • Useful for research on respiratory diseases
  • Inhibits histamine-induced contraction of isolated guinea pig trachea (in vitro)
  • Reduces lipopolysaccharide-induced tumor necrosis factor concentrations (in vivo)
  • Reduces primed stimulation of alveolar macrophages (in vivo)
Catalog No. 50-004-32930 Supplier Medchemexpress LLC Supplier No. HYA002710MM/1ML
Fenspiride hydrochloride is an orally active, non-steroidal anti-inflammatory agent that acts as an antagonist of the H1-histamine receptor. It inhibits phosphodiesterase 3 (PDE3), phosphodiesterase 4 (PDE4), and phosphodiesterase 5 (PDE5) activities. This compound can be used for research related to respiratory diseases.
  • Antagonist of the H1-histamine receptor
  • Inhibits phosphodiesterase 3, phosphodiesterase 4, and phosphodiesterase 5 activities
  • Useful for research on respiratory diseases
  • Inhibits histamine-induced contraction of isolated guinea pig trachea (in vitro)
  • Reduces lipopolysaccharide-induced tumor necrosis factor concentrations (in vivo)
  • Reduces primed stimulation of alveolar macrophages (in vivo)